Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011247.2 | NP_035377.2 | 1 | 527 | ||
NM_175023.3 | NP_778188.1 | 1 | 527 | |||
GenBank mRNA | AK045635.1 | - | 1 | 272 | ||
AK081261.1 | - | 1 | 391 | |||
BC025874.1 | - | 1 | 164 | |||
BE994197.1 | - | 1 | 527 |