Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011251.3 | NP_035381.2 | 13, 14 | 2589 | ||
NM_029169.3 | NP_083445.1 | 13, 14 | 2860 | |||
GenBank mRNA | AJ006486.2 | - | 13, 14 | 2468 | ||
AK013915.1 | - | 4, 5 | 282 | |||
AK032150.1 | - | 8, 9 | 841 | |||
AK077899.1 | - | 13, 14 | 2767 | |||
AK145612.1 | - | 13, 14 | 2749 | |||
AK164432.1 | - | 8, 9 | 946 | |||
AK220446.1 | - | 13, 14 | 2738 | |||
BC026129.1 | - | 13, 14 | 2614 | 3982861 | ||
BC026130.1 | - | 13, 14 | 2763 |