Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbpj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080927.2 | NP_001074396.1 | 3 | 356 | ||
NM_001080928.1 | NP_001074397.1 | 3 | 242 | |||
NM_001277116.1 | NP_001264045.1 | 5 | 438 | |||
NM_009035.5 | NP_033061.3 | 3 | 258 | |||
GenBank mRNA | AK028363.1 | - | 5 | 416 | ||
AK080359.1 | - | 3 | 151 | |||
AK090006.1 | - | 3 | 144 | |||
AK132145.1 | - | 3 | 200 | |||
AK139087.1 | - | 3 | 143 | |||
AK145187.1 | - | 3 | 150 | |||
AK152746.1 | - | 3 | 259 | |||
AK153163.1 | - | 4 | 146 | |||
BB839243.1 | - | 3 | 243 | |||
BC035299.1 | - | 1 | 44 | |||
BC051387.1 | - | 3 | 187 | 4460290 | ||
BY155397.1 | - | 3 | 385 | |||
BY716845.1 | - | 3 | 329 | |||
CD540070.1 | - | 3 | 196 | |||
S63463.1 | - | 3 | 259 | |||
X17459.1 | - | 3 | 254 |