Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009047.5 | NP_033073.1 | 2 | 428 | ||
GenBank mRNA | AK053133.1 | - | 2 | 429 | ||
AK084460.1 | - | 2 | 436 | |||
BC150768.1 | - | 1 | 280 | 9087679 | ||
U91601.1 | - | 2 | 406 |