Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009047.5 | NP_033073.1 | 3, 4 | 649 | ||
GenBank mRNA | AK053133.1 | - | 3, 4 | 650 | ||
AK084460.1 | - | 3, 4 | 657 | |||
BC150768.1 | - | 2, 3 | 501 | 9087679 | ||
U91601.1 | - | 3, 4 | 627 |