Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020022.2 | NP_064406.1 | 10, 11 | 1035 | ||
GenBank mRNA | AK011353.1 | - | 10, 11 | 951 | ||
AK011649.1 | - | 10, 11 | 948 | |||
AK075997.1 | - | 10, 11 | 947 | |||
AK154681.1 | - | 10, 11 | 989 | |||
BC004812.1 | - | 7, 8 | 707 | |||
BC023028.1 | - | 10, 11 | 956 | 5374851 | ||
BC099370.1 | - | 10, 11 | 999 | 30937690 |