Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016846.3 | NP_058542.2 | 5, 6 | 710 | ||
GenBank mRNA | AK144218.1 | - | 5, 6 | 785 | ||
AK169109.1 | - | 5, 6 | 762 | |||
AK169180.1 | - | 5, 6 | 763 | |||
BC017678.1 | - | 5, 6 | 710 | 4241244 | ||
U14103.1 | - | 5, 6 | 768 |