Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rlbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173483.1 | NP_001166954.1 | 8 | 1217 | ||
NM_020599.2 | NP_065624.1 | 8 | 1299 | |||
GenBank mRNA | AK014227.1 | - | 8 | 990 | ||
AK140622.1 | - | 8 | 1218 | |||
BC016268.1 | - | 7 | 892 | 4481973 |