Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011273.2 | NP_035403.1 | 10 | 1495 | ||
GenBank mRNA | AF114753.1 | - | 10 | 1380 | ||
AF131096.1 | - | 10 | 1285 | |||
AF131098.1 | - | 10 | 1285 | |||
AF131102.1 | - | 10 | 1285 | |||
AF198104.1 | - | 10 | 1285 | |||
AK087140.1 | - | 1 | 671 | |||
AK141660.1 | - | 10 | 1467 | |||
AK147403.1 | - | 10 | 1363 | |||
BC153872.1 | - | 10 | 1371 | 40110725 | ||
HQ022845.1 | - | 10 | 1285 | |||
HQ022853.1 | - | 10 | 1280 | |||
HQ323678.1 | - | 1 | 136 | |||
HQ323679.1 | - | 10 | 1285 | |||
HQ323680.1 | - | 10 | 1285 |
Set Membership: |
G-protein coupled receptor |