Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF169300.1 | - | 3 | 371 | ||
AK019171.1 | - | 3 | 386 | |||
AK019656.1 | - | 3 | 423 | |||
AK090162.1 | - | 3 | 409 | |||
AK133924.1 | - | 3 | 471 | |||
AK145596.1 | - | 3 | 348 | |||
AK147057.1 | - | 3 | 331 | |||
AK151116.1 | - | 3 | 379 | |||
AK158987.1 | - | 3 | 388 | |||
AK159296.1 | - | 3 | 386 | |||
AK159778.1 | - | 3 | 380 | |||
AK159949.1 | - | 3 | 428 | |||
AK207872.1 | - | 3 | 309 | |||
BC003282.1 | - | 3 | 350 | 3582133 | ||
BY275961.1 | - | 3 | 361 | |||
U95141.1 | - | 1 | 61 |