Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF169300.1 | - | 4 | 454 | ||
AK019171.1 | - | 4 | 469 | |||
AK019656.1 | - | 4 | 505 | |||
AK090162.1 | - | 4 | 492 | |||
AK133924.1 | - | 4 | 554 | |||
AK145596.1 | - | 4 | 431 | |||
AK147057.1 | - | 4 | 414 | |||
AK151116.1 | - | 4 | 462 | |||
AK158987.1 | - | 4 | 471 | |||
AK159296.1 | - | 4 | 469 | |||
AK159778.1 | - | 4 | 463 | |||
AK159949.1 | - | 4 | 511 | |||
BC003282.1 | - | 4 | 433 | 3582133 | ||
U95141.1 | - | 2 | 144 |