Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009086.2 | NP_033112.2 | 15 | 3475 | ||
GenBank mRNA | AK083732.1 | - | 15 | 3458 | ||
AK143501.1 | - | 15 | 3412 | |||
AK144616.1 | - | 15 | 3462 | |||
AK156712.1 | - | 15 | 3465 | |||
BC060656.1 | - | 15 | 3405 | 30355130 | ||
U58280.1 | - | 15 | 3439 |