Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ryr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009109.2 | NP_033135.2 | 7 | 711 | ||
GenBank mRNA | AK143310.1 | - | 5 | 1380 | ||
AY268935.1 | - | 7 | 711 |