Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ryr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177652.2 | NP_808320.2 | 17 | 1902 | ||
GenBank mRNA | AK134079.1 | - | 2 | 160 |