Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009112.2 | NP_033138.1 | 3 | 357 | ||
GenBank mRNA | AK149077.1 | - | 3 | 343 | ||
AK161647.1 | - | 3 | 341 | |||
AK170919.1 | - | 3 | 337 | |||
BC025044.1 | - | 3 | 288 | 4949465 | ||
BU559648.1 | - | 360 | ||||
M16465.1 | - | 3 | 310 |