Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013650.2 | NP_038678.1 | 2 | 135 | ||
GenBank mRNA | AK157837.1 | - | 2 | 136 | ||
BC078629.1 | - | 2 | 121 | 864657 | ||
M83218.1 | - | 1 | 97 | |||
S57123.1 | - | 2 | 122 | |||
Y09039.1 | - | 1 | 85 |