Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011333.3 | NP_035463.1 | 1, 2 | 149 | ||
GenBank mRNA | AF065929.1 | - | 1, 2 | 87 | ||
AF065930.1 | - | 1, 2 | 87 | |||
AF065931.1 | - | 1, 2 | 87 | |||
AF065932.1 | - | 1, 2 | 87 | |||
AF065933.1 | - | 1, 2 | 87 | |||
AK132590.1 | - | 1, 2 | 148 | |||
AK150937.1 | - | 1, 2 | 150 | |||
AK151789.1 | - | 1, 2 | 148 | |||
AK153443.1 | - | 1, 2 | 148 | |||
AK153468.1 | - | 1, 2 | 150 | |||
AK153520.1 | - | 1, 2 | 150 | |||
BC055070.1 | - | 1, 2 | 127 | |||
BC145867.1 | - | 1, 2 | 116 | 40131170 | ||
BC145869.1 | - | 1, 2 | 116 | 40131172 | ||
CT010187.1 | - | 1, 2 | 64 |