Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159738.1 | NP_001153210.1 | 2, 3 | 243 | ||
NM_016960.2 | NP_058656.1 | 2, 3 | 246 | |||
GenBank mRNA | AB015136.1 | - | 2, 3 | 230 | ||
AF053313.1 | - | 2, 3 | 222 | |||
AF099052.1 | - | 2, 3 | 216 | |||
AJ222694.1 | - | 2, 3 | 206 | |||
AK079862.1 | - | 2, 3 | 244 | |||
BC028504.1 | - | 2, 3 | 216 | 1380543 | ||
BX525886.1 | - | 2, 3 | 224 | |||
BY026108.1 | - | 2, 3 | 243 |