Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sord | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146126.4 | NP_666238.1 | 6 | 640 | ||
GenBank mRNA | AK004692.1 | - | 6 | 664 | ||
AK015059.1 | - | 6 | 641 | |||
AK152169.1 | - | 6 | 689 | |||
AK166988.1 | - | 6 | 640 | |||
AK166996.1 | - | 6 | 669 | |||
AK219035.1 | - | 4 | 369 | |||
BC024124.1 | - | 6 | 655 | 5099395 | ||
BC030875.1 | - | 6 | 629 | 4237164 | ||
BC039957.1 | - | 6 | 640 | |||
BC092291.1 | - | 6 | 643 | 5097505 | ||
U27014.1 | - | 640 |