Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252332.1 | NP_001239261.1 | 5 | 920 | ||
NM_001252333.1 | NP_001239262.1 | 5 | 914 | |||
NM_033144.2 | NP_149156.1 | 5 | 920 | |||
GenBank mRNA | AB093215.1 | - | 5 | 770 | ||
AF179996.1 | - | 4 | 616 | |||
AK144178.1 | - | 5 | 756 | |||
BC037707.1 | - | 5 | 708 | |||
BC049819.1 | - | 5 | 735 | |||
BC059248.1 | - | 5 | 856 | 6400429 |