Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sez6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291225.1 | NP_001278154.1 | 10 | 2302 | ||
NM_021286.4 | NP_067261.2 | 10 | 2302 | |||
GenBank mRNA | AB206791.1 | - | 10 | 2238 | ||
AK147705.1 | - | 8 | 2544 | |||
AK161576.1 | - | 5 | 712 | |||
AK220513.1 | - | 9 | 2932 | |||
BC053011.1 | - | 10 | 2198 | 5698186 | ||
BC055345.1 | - | 8 | 2625 | 6833344 | ||
D29763.1 | - | 10 | 2129 | |||
D64009.1 | - | 10 | 2102 | |||
D64010.1 | - | 9 | 2315 |