Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011355.1 | NP_035485.1 | 5 | 715 | ||
GenBank mRNA | AK142246.1 | - | 5 | 732 | ||
AK153524.1 | - | 5 | 746 | |||
AK154945.1 | - | 5 | 767 | |||
AK155085.1 | - | 5 | 751 | |||
BC003815.1 | - | 5 | 633 | 3600260 | ||
L03215.1 | - | 5 | 612 | |||
M32370.1 | - | 5 | 695 | |||
M38252.1 | - | 5 | 715 | |||
X17463.1 | - | 5 | 1403 |