Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sftpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023134.4 | NP_075623.2 | 3 | 131 | ||
GenBank mRNA | AK004620.1 | - | 3 | 132 | ||
AK004788.1 | - | 3 | 132 | |||
AK011333.1 | - | 2 | 105 | |||
AK075702.1 | - | 3 | 133 | |||
BC061096.1 | - | 3 | 131 | 30300708 |