Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sftpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023134.4 | NP_075623.2 | 6 | 787 | ||
GenBank mRNA | AK004620.1 | - | 6 | 788 | ||
AK004788.1 | - | 6 | 788 | |||
AK011333.1 | - | 5 | 761 | |||
AK075702.1 | - | 6 | 794 | |||
BC061096.1 | - | 6 | 604 | 30300708 |