Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009161.4 | NP_033187.1 | 10 | 1329 | ||
GenBank mRNA | AB024920.1 | - | 10 | 1329 | ||
AF019564.1 | - | 9 | 1076 | |||
AK009015.1 | - | 9 | 1087 | |||
AK075915.1 | - | 9 | 1101 | |||
BC060287.1 | - | 9 | 1081 |