Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159745.1 | NP_001153217.1 | 4 | 1048 | ||
NM_009183.2 | NP_033209.1 | 4 | 1048 | |||
GenBank mRNA | AJ223956.1 | - | 4 | 1039 | ||
AK003690.1 | - | 4 | 990 | |||
AK041723.1 | - | 4 | 981 | |||
BC060112.1 | - | 4 | 1025 | 6405078 | ||
X86000.1 | - | 1048 |