Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Six6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011384.4 | NP_035514.1 | 1 | 387 | ||
GenBank mRNA | AF050130.2 | - | 1 | 422 | ||
AF135267.1 | - | 1 | 438 | |||
AJ011787.1 | - | 1 | 276 | |||
AK017544.1 | - | 1 | 451 | |||
AK029309.1 | - | 1 | 388 | |||
BC138838.1 | - | 1 | 360 | 8861860 | ||
BC138839.1 | - | 1 | 360 | 8861861 |