Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253804.1 | NP_001240733.1 | 7 | 829 | ||
NM_009195.3 | NP_033221.1 | 7 | 823 | |||
GenBank mRNA | AF047339.1 | - | 7 | 747 | ||
AF121118.1 | - | 7 | 814 | |||
AK149969.1 | - | 7 | 812 | |||
AK151717.1 | - | 7 | 784 | |||
AK152762.1 | - | 7 | 801 | |||
AK153245.1 | - | 7 | 801 | |||
AK159274.1 | - | 7 | 811 | |||
AK159331.1 | - | 7 | 806 | |||
AK159483.1 | - | 7 | 779 | |||
AK159537.1 | - | 7 | 806 | |||
AK160034.1 | - | 7 | 772 | |||
AK160072.1 | - | 7 | 806 | |||
AK164479.1 | - | 7 | 788 | |||
AK165874.1 | - | 7 | 801 | |||
AK166304.1 | - | 7 | 782 | |||
AK167371.1 | - | 7 | 820 | |||
AK167444.1 | - | 7 | 820 | |||
AK169044.1 | - | 7 | 778 | |||
AK213204.1 | - | 1 | 59 | |||
BC066872.1 | - | 7 | 753 | 30436525 |