Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019810.4 | NP_062784.3 | 9 | 1240 | ||
GenBank mRNA | AF163846.1 | - | 9 | 1002 | ||
AF208031.1 | - | 9 | 1240 | |||
AK033512.1 | - | 9 | 1226 | |||
AK085602.1 | - | 9 | 1031 | |||
BC003845.1 | - | 9 | 1220 | 3497611 | ||
U27269.1 | - | 7 | 810 |