Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301671.1 | NP_001288600.1 | 4 | 669 | ||
NM_001301672.1 | NP_001288601.1 | 4 | 501 | |||
NM_001301673.1 | NP_001288602.1 | 4 | 552 | |||
NM_019929.4 | NP_064313.1 | 4 | 543 | |||
GenBank mRNA | AA105565.1 | - | 3 | 273 | ||
AF063847.1 | - | 4 | 370 | |||
AK012742.1 | - | 4 | 418 | |||
AK013019.1 | - | 4 | 412 | |||
AK148306.1 | - | 4 | 407 | |||
AK150063.1 | - | 4 | 416 | |||
AK150200.1 | - | 4 | 416 | |||
AK160169.1 | - | 4 | 415 | |||
AK162035.1 | - | 4 | 416 | |||
AK167207.1 | - | 4 | 384 | |||
AK168988.1 | - | 4 | 407 | |||
AK170896.1 | - | 4 | 601 | |||
AK171169.1 | - | 4 | 601 | |||
BC115488.1 | - | 4 | 350 | 40100722 | ||
BC115489.1 | - | 4 | 350 | 40100723 | ||
CK619326.1 | - | 5 | 511 |