Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011427.2 | NP_035557.1 | 2 | 459 | ||
GenBank mRNA | AK144478.1 | - | 2 | 482 | ||
BC034857.1 | - | 2 | 459 | 5121591 | ||
CT010204.1 | - | 2 | 396 | |||
CT010370.1 | - | 2 | 396 | |||
M95604.1 | - | 2 | 410 | |||
X67253.1 | - | 2 | 416 |