Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009223.3 | NP_033249.1 | 2 | 364 | ||
GenBank mRNA | AK149685.1 | - | 2 | 313 | ||
BC006961.1 | - | 2 | 332 | 3707801 | ||
BC017685.1 | - | 2 | 281 | 4241981 | ||
BC038475.1 | - | 1 | 405 | 3157762 | ||
BC063027.1 | - | 2 | 301 | 5343626 | ||
BC087932.1 | - | 309 | 6706156 | |||
BY277718.1 | - | 2 | 365 | |||
CJ166486.1 | - | 2 | 316 |