Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005045.2 | - | 10 | 2197 | ||
AK011564.1 | - | 9 | 793 | |||
AK014311.2 | - | 9 | 834 | |||
AK077425.1 | - | 9 | 3219 | |||
AK133115.1 | - | 9 | 838 | |||
AK146729.1 | - | 9 | 838 | |||
AK148013.1 | - | 9 | 865 | |||
BC002169.1 | - | 2 | 102 | |||
BC049128.1 | - | 9 | 3169 | |||
BC058513.1 | - | 2 | 343 | |||
BC094067.1 | - | 2 | 494 | |||
BC131994.1 | - | 9 | 730 | 40130271 | ||
BC132020.1 | - | 9 | 730 | 40130297 |