Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sntb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009229.4 | NP_033255.1 | 4, 5 | 1145 | ||
GenBank mRNA | AK040423.1 | - | 4, 5 | 1146 | ||
AK080059.1 | - | 4, 5 | 1142 | |||
AK131687.1 | - | 4, 5 | 1170 | |||
AK165009.1 | - | 4, 5 | 1147 | |||
AK165900.1 | - | 4, 5 | 1122 | |||
BC030352.1 | - | 3, 4 | 586 | |||
BC038704.1 | - | 3, 4 | 517 | |||
BC051571.1 | - | 1, 2 | 39 | |||
BC138850.1 | - | 4, 5 | 1144 | 8861872 | ||
BC145442.1 | - | 4, 5 | 1144 | 9054000 | ||
U00678.1 | - | 4, 5 | 1144 |