Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sorl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011436.3 | NP_035566.2 | 20 | 3037 | ||
GenBank mRNA | AF031816.1 | - | 2315 | |||
AK137635.1 | - | 8 | 1152 | |||
AK147303.1 | - | 20 | 3038 |
Gene Symbol: LOC102640140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037519.1 | - | 574 |