Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bptf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176850.2 | NP_789820.2 | 11 | 3796 | ||
GenBank mRNA | GU189417.1 | - | 11 | 3349 |