Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hectd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144788.2 | NP_659037.2 | 9, 10 | 1811 | ||
GenBank mRNA | AK042387.1 | - | 9, 10 | 1585 | ||
AK153674.1 | - | 9, 10 | 1628 | |||
AK169603.1 | - | 9, 10 | 1604 | |||
AK172326.1 | - | 9, 10 | 1812 | |||
AK213219.1 | - | 1, 2 | 99 | |||
BC031446.1 | - | 9, 10 | 1551 | |||
BC048868.1 | - | 5, 6 | 640 |