Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009258.5 | NP_033284.1 | 3 | 205 | ||
GenBank mRNA | AK007841.1 | - | 3 | 166 | ||
AK007985.1 | - | 3 | 164 | |||
AV048854.1 | - | 3 | 95 | |||
BC086887.1 | - | 3 | 164 | 6775994 | ||
CA481615.1 | - | 3 | 242 | |||
X06342.1 | - | 3 | 162 |