Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016907.3 | NP_058603.2 | 4, 5 | 924 | ||
GenBank mRNA | AF099018.1 | - | 4, 5 | 879 | ||
AK017342.1 | - | 3, 4 | 806 | |||
BC005769.1 | - | 3, 4 | 811 | 2650110 |