Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011465.4 | NP_035595.2 | 24, 25 | 3630 | ||
GenBank mRNA | AF093576.1 | - | 24, 25 | 3538 |