Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009260.2 | NP_033286.2 | 23, 24 | 5379 | ||
NM_175836.2 | NP_787030.2 | 23, 24 | 5039 | |||
GenBank mRNA | AF017112.1 | - | 23, 24 | 5286 | ||
AK161119.1 | - | 2, 3 | 162 | |||
AK220456.1 | - | 24, 25 | 5241 | |||
M74773.1 | - | 23, 24 | 5359 |