Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009267.2 | NP_033293.2 | 3 | 235 | ||
GenBank mRNA | AK006124.1 | - | 3 | 233 | ||
BC030164.1 | - | 3 | 228 | |||
BC106151.1 | - | 3 | 219 | 4923478 | ||
CK616337.1 | - | 2 | 413 | |||
CK616968.1 | - | 3 | 236 | |||
M33974.1 | - | 3 | 231 | |||
X00349.1 | - | 3 | 232 |
Gene Symbol: Mucl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC091766.1 | - | 194 | 4190032 |