Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam228b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175431.4 | NP_780640.2 | 7 | 1050 | ||
GenBank mRNA | AK044133.1 | - | 6 | 1137 | ||
AK050447.1 | - | 7 | 1017 | |||
BC100429.1 | - | 7 | 829 | 30917787 | ||
BC144941.1 | - | 8 | 929 | |||
BC145648.1 | - | 7 | 781 | 40130951 |