Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map6d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198599.2 | NP_941001.2 | 1, 2 | 423 | ||
GenBank mRNA | AK079603.1 | - | 1, 2 | 386 | ||
AK163656.1 | - | 1, 2 | 387 | |||
BB653005.1 | - | 1, 2 | 424 | |||
BC116220.1 | - | 1, 2 | 386 | 40091320 | ||
BC116221.1 | - | 1, 2 | 386 | 40091321 | ||
BE979311.1 | - | 1, 2 | 97 |