Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009275.4 | NP_033301.1 | 5 | 495 | ||
GenBank mRNA | AK004854.1 | - | 5 | 528 | ||
AK007859.1 | - | 5 | 501 | |||
AK008383.1 | - | 5 | 538 | |||
AK015469.1 | - | 5 | 560 | |||
AK030437.1 | - | 5 | 496 | |||
AK075583.1 | - | 5 | 498 | |||
AK160600.1 | - | 4 | 1729 | |||
AK167039.1 | - | 5 | 501 | |||
AK179126.1 | - | 1 | 48 | |||
AK179649.1 | - | 1 | 48 | |||
AK181904.1 | - | 1 | 48 | |||
AK186925.1 | - | 1 | 48 | |||
AK216962.1 | - | 1 | 48 | |||
AK219957.1 | - | 4 | 263 | |||
BC003798.1 | - | 5 | 516 | 3490766 | ||
U17343.1 | - | 5 | 509 |