Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011484.2 | NP_035614.1 | 14 | 1583 | ||
GenBank mRNA | AK144987.1 | - | 14 | 1600 | ||
AK157767.1 | - | 14 | 1530 | |||
AK162431.1 | - | 14 | 1530 | |||
AK163720.1 | - | 14 | 1533 | |||
AK163922.1 | - | 14 | 1520 | |||
AK164191.1 | - | 6 | 515 | |||
AK169170.1 | - | 14 | 1590 | |||
BC044666.1 | - | 14 | 1583 | 4950569 | ||
BC055326.1 | - | 14 | 1529 | 6416552 | ||
U43900.1 | - | 14 | 1440 |