Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011486.4 | NP_035616.1 | 18 | 1916 | ||
NM_213659.2 | NP_998824.1 | 18 | 1916 | |||
NM_213660.2 | NP_998825.1 | 18 | 1916 | |||
GenBank mRNA | AK145486.1 | - | 18 | 1772 | ||
AK153005.1 | - | 18 | 1828 | |||
AK153472.1 | - | 18 | 1828 | |||
AK193746.1 | - | 2 | 86 | |||
AY299489.1 | - | 17 | 1643 | |||
AY299490.1 | - | 17 | 1643 | |||
BC003806.1 | - | 18 | 1804 | 3593588 | ||
BC019168.1 | - | 18 | 1820 | 4923137 | ||
BC037688.1 | - | 2 | 222 | |||
L29278.1 | - | 18 | 1895 | |||
U06922.1 | - | 18 | 1705 | |||
U08378.1 | - | 18 | 1783 | |||
U30709.1 | - | 18 | 1809 |