Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Supt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009297.2 | NP_033323.2 | 17 | 2280 | ||
GenBank mRNA | AK129069.1 | - | 5 | 603 | ||
AK170533.1 | - | 17 | 2279 | |||
BC072657.1 | - | 17 | 2200 | 30536338 | ||
U40375.1 | - | 17 | 2278 |