Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svs6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013679.2 | NP_038707.2 | 2, 3 | 138 | ||
GenBank mRNA | AK020303.1 | - | 2, 3 | 140 | ||
BC119247.1 | - | 2, 3 | 142 | 8733996 | ||
BC119249.1 | - | 2, 3 | 142 | 8733998 | ||
L11393.1 | - | 2, 3 | 130 | |||
S65753.1 | - | 2, 3 | 130 |