Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009305.2 | NP_033331.2 | 2, 3 | 220 | ||
GenBank mRNA | AK034291.1 | - | 2, 3 | 163 | ||
AK043756.1 | - | 2, 3 | 117 | |||
AK090259.1 | - | 2, 3 | 117 | |||
BC014823.1 | - | 2, 3 | 90 | 4507116 | ||
BC052354.1 | - | 2, 3 | 114 | |||
BC115644.1 | - | 1, 2 | 73 | |||
BC115645.1 | - | 1, 2 | 72 | |||
BY239748.1 | - | 2, 3 | 220 | |||
X95818.1 | - | 2, 3 | 89 |